Assay Details
| Gene Symbol: Hinfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856776.2 | XP_004856833.1 | 4 - 5 | 1009 | 62 | |
| XM_004856777.2 | XP_004856834.1 | 4 - 5 | 620 | 62 | ||
| XM_021237760.1 | XP_021093419.1 | 5 - 6 | 1139 | 62 | ||
More Information
Set Membership: |
|

Hinfp
Genomic Map