Assay Details
| Gene Symbol: Rsrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843900.3 | XP_004843957.1 | 6 - 7 | 899 | 68 | |
| XM_004843901.3 | XP_004843958.1 | 6 - 7 | 805 | 68 | ||
| XM_021257572.1 | XP_021113231.1 | 6 - 7 | 921 | 68 | ||
More Information
Set Membership: |
|

Rsrc2
Genomic Map