Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851698.3 | XP_004851755.1 | 6 - 7 | 905 | 63 | |
| XM_013074628.2 | XP_012930082.1 | 6 - 7 | 905 | 63 | ||
| XM_021264048.1 | XP_021119707.1 | 6 - 7 | 1008 | 63 | ||
More Information
Set Membership: |
|

Deaf1
Genomic Map