Assay Details
| Gene Symbol: Rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866904.3 | XP_004866961.1 | 4 - 5 | 505 | 61 | |
| XM_021245771.1 | XP_021101430.1 | 4 - 5 | 600 | 61 | ||
More Information
Set Membership: |
|

Rras
Genomic Map