Assay Details
| Gene Symbol: LOC101708260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240277.1 | XP_021095936.1 | 10 - 11 | 1408 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101708260| Gene Symbol: LOC101708260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240277.1 | XP_021095936.1 | 10 - 11 | 1408 | 74 | |
Set Membership: |
|