Assay Details
| Gene Symbol: LOC101719219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072683.2 | XP_012928137.1 | 11 - 12 | 1174 | 98 | |
| XM_013072684.2 | XP_012928138.1 | 12 - 13 | 2356 | 98 | ||
| XM_013072686.2 | XP_012928140.1 | 11 - 12 | 2259 | 98 | ||
| XM_021259205.1 | XP_021114864.1 | 15 - 16 | 1627 | 98 | ||
| XM_021259206.1 | XP_021114865.1 | 14 - 15 | 1599 | 98 | ||
| XM_021259208.1 | XP_021114867.1 | 14 - 15 | 1463 | 98 | ||
| XM_021259209.1 | XP_021114868.1 | 14 - 15 | 1540 | 98 | ||
| XM_021259210.1 | XP_021114869.1 | 16 - 17 | 1684 | 98 | ||
| XM_021259211.1 | XP_021114870.1 | 13 - 14 | 1401 | 98 | ||
| XM_021259212.1 | XP_021114871.1 | 11 - 12 | 1127 | 98 | ||
| XM_021259213.1 | XP_021114872.1 | 4 - 5 | 1734 | 98 | ||
More Information
Set Membership: |
|

LOC101719219
Genomic Map