Assay Details
| Gene Symbol: LOC101719219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072683.2 | XP_012928137.1 | 8 - 9 | 801 | 73 | |
| XM_013072684.2 | XP_012928138.1 | 9 - 10 | 1983 | 73 | ||
| XM_013072686.2 | XP_012928140.1 | 8 - 9 | 1886 | 73 | ||
| XM_021259205.1 | XP_021114864.1 | 12 - 13 | 1254 | 73 | ||
| XM_021259206.1 | XP_021114865.1 | 11 - 12 | 1226 | 73 | ||
| XM_021259207.1 | XP_021114866.1 | 11 - 12 | 1090 | 73 | ||
| XM_021259208.1 | XP_021114867.1 | 11 - 12 | 1090 | 73 | ||
| XM_021259209.1 | XP_021114868.1 | 11 - 12 | 1167 | 73 | ||
| XM_021259210.1 | XP_021114869.1 | 13 - 14 | 1311 | 73 | ||
| XM_021259211.1 | XP_021114870.1 | 10 - 11 | 1028 | 73 | ||
| XM_021259212.1 | XP_021114871.1 | 8 - 9 | 754 | 73 | ||
More Information
Set Membership: |
|

LOC101719219
Genomic Map