Assay Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869611.2 | XP_004869668.1 | 8 - 9 | 2271 | 68 | |
| XM_004869612.1 | XP_004869669.1 | 8 - 9 | 1410 | 68 | ||
| XM_004869613.2 | XP_004869670.1 | 8 - 9 | 1414 | 68 | ||
| XM_013067411.1 | XP_012922865.1 | 7 - 8 | 1302 | 68 | ||
| XM_013067413.1 | XP_012922867.1 | 7 - 8 | 1233 | 68 | ||
| XM_021248211.1 | XP_021103870.1 | 9 - 10 | 2405 | 68 | ||
More Information
Set Membership: |
|

Oma1
Genomic Map