Assay Details
| Gene Symbol: Mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858825.2 | XP_004858882.1 | 2 - 3 | 161 | 63 | |
| XM_004858826.3 | XP_004858883.1 | 2 - 3 | 229 | 63 | ||
| XM_004858827.3 | XP_004858884.1 | 3 - 4 | 236 | 63 | ||
| XM_004858830.2 | XP_004858887.1 | 2 - 3 | 161 | 63 | ||
| XM_013077555.2 | XP_012933009.1 | 3 - 4 | 175 | 63 | ||
| XM_021239721.1 | XP_021095380.1 | 2 - 3 | 93 | 63 | ||
More Information
Set Membership: |
|

Mag
Genomic Map