Assay Details
| Gene Symbol: Bpnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867398.3 | XP_004867455.1 | 5 - 6 | 551 | 63 | |
| XM_013066534.2 | XP_012921988.1 | 4 - 5 | 443 | 63 | ||
More Information
Set Membership: |
|

Bpnt1
Genomic Map