Assay Details
Gene Symbol: LOC101703804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864580.2 | XP_004864637.1 | 6 - 7 | 1148 | 92 | |
XM_004864582.2 | XP_004864639.1 | 6 - 7 | 1117 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
