Assay Details
| Gene Symbol: Ahsa2p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246280.1 | XP_021101939.1 | 4 - 5 | 577 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahsa2p| Gene Symbol: Ahsa2p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246280.1 | XP_021101939.1 | 4 - 5 | 577 | 80 | |
Set Membership: |
|