Assay Details
| Gene Symbol: LOC101697932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839953.3 | XP_004840010.1 | 6 - 7 | 859 | 93 | |
| XM_004839954.3 | XP_004840011.1 | 2 - 3 | 448 | 93 | ||
| XM_004839955.3 | XP_004840012.1 | 2 - 3 | 273 | 93 | ||
| XM_004839956.3 | XP_004840013.1 | 2 - 3 | 310 | 93 | ||
More Information
Set Membership: |
|

LOC101697932
Genomic Map