Assay Details
| Gene Symbol: Plekha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839485.2 | XP_004839542.1 | 1 - 2 | 407 | 64 | |
| XM_013070275.2 | XP_012925729.1 | 1 - 2 | 408 | 64 | ||
More Information
Set Membership: |
|

Plekha8
Genomic Map