Assay Details
| Gene Symbol: LOC101709796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840075.3 | XP_004840132.1 | 6 - 7 | 1304 | 94 | |
| XM_021254425.1 | XP_021110084.1 | 5 - 6 | 1223 | 94 | ||
| XM_021254426.1 | XP_021110085.1 | 6 - 7 | 856 | 94 | ||
More Information
Set Membership: |
|

LOC101709796
Genomic Map