Assay Details
| Gene Symbol: Kirrel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239853.1 | XP_021095512.1 | 8 - 9 | 1530 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kirrel1| Gene Symbol: Kirrel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239853.1 | XP_021095512.1 | 8 - 9 | 1530 | 106 | |
Set Membership: |
|