Assay Details
| Gene Symbol: Folh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868200.2 | XP_004868257.1 | 11 - 12 | 1365 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Folh1b| Gene Symbol: Folh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868200.2 | XP_004868257.1 | 11 - 12 | 1365 | 83 | |
Set Membership: |
|