Assay Details
| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848428.2 | XP_004848485.1 | 3 - 4 | 2959 | 56 | |
| XM_004848430.3 | XP_004848487.1 | 3 - 4 | 2880 | 56 | ||
| XM_021261338.1 | XP_021116997.1 | 3 - 4 | 2896 | 56 | ||
More Information
Set Membership: |
|

Extl3
Genomic Map