Assay Details
| Gene Symbol: Igf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844842.3 | XP_004844899.1 | 3 - 4 | 671 | 107 | |
| XM_013072263.2 | XP_012927717.1 | 3 - 4 | 758 | 107 | ||
More Information
Set Membership: |
|

Igf1
Genomic Map