Assay Details
| Gene Symbol: Caspase-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874015.3 | XP_004874072.1 | 4 - 5 | 683 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caspase-12| Gene Symbol: Caspase-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874015.3 | XP_004874072.1 | 4 - 5 | 683 | 62 | |
Set Membership: |
|