Assay Details
| Gene Symbol: Spock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846235.3 | XP_004846292.1 | 9 - 10 | 1233 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spock2| Gene Symbol: Spock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846235.3 | XP_004846292.1 | 9 - 10 | 1233 | 68 | |
Set Membership: |
|