Assay Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874183.3 | XP_004874240.1 | 1 - 2 | 1601 | 88 | |
| XM_021251682.1 | XP_021107341.1 | 1 - 2 | 1095 | 88 | ||
More Information
Set Membership: |
|

Hipk4
Genomic Map