Assay Details
| Gene Symbol: Drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852351.3 | XP_004852408.1 | 5 - 6 | 598 | 81 | |
| XM_013075026.2 | XP_012930480.1 | 5 - 6 | 619 | 81 | ||
| XM_021264563.1 | XP_021120222.1 | 5 - 6 | 619 | 81 | ||
| XM_021264564.1 | XP_021120223.1 | 5 - 6 | 598 | 81 | ||
| XM_021264565.1 | XP_021120224.1 | 4 - 5 | 542 | 81 | ||
More Information
Set Membership: |
|

Drap1
Genomic Map