Assay Details
| Gene Symbol: Atp6v0d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843032.3 | XP_004843089.1 | 7 - 8 | 1013 | 61 | |
| XM_021256862.1 | XP_021112521.1 | 7 - 8 | 1062 | 61 | ||
| XM_021256863.1 | XP_021112522.1 | 7 - 8 | 1062 | 61 | ||
| XM_021256864.1 | XP_021112523.1 | 7 - 8 | 1062 | 61 | ||
More Information
Set Membership: |
|

Atp6v0d1
Genomic Map