Assay Details
| Gene Symbol: Ap1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021244203.1 | XP_021099862.1 | 2 - 3 | 559 | 103 | |
| XM_021244204.1 | XP_021099863.1 | 2 - 3 | 326 | 103 | ||
| XM_021244205.1 | XP_021099864.1 | 2 - 3 | 541 | 103 | ||
More Information
Set Membership: |
|

Ap1s3
Genomic Map