Assay Details
| Gene Symbol: Dusp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845864.3 | XP_004845921.1 | 5 - 6 | 1142 | 68 | |
| XM_004845865.3 | XP_004845922.1 | 5 - 6 | 1416 | 68 | ||
| XM_004845867.3 | XP_004845924.1 | 5 - 6 | 1903 | 68 | ||
More Information
Set Membership: |
|

Dusp16
Genomic Map