Assay Details
| Gene Symbol: Zfand2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065461.2 | XP_012920915.1 | 7 - 8 | 678 | 58 | |
| XM_013065462.2 | XP_012920916.1 | 7 - 8 | 655 | 58 | ||
| XM_021244247.1 | XP_021099906.1 | 7 - 8 | 699 | 58 | ||
| XM_021244248.1 | XP_021099907.1 | 7 - 8 | 676 | 58 | ||
More Information
Set Membership: |
|

Zfand2b
Genomic Map