Assay Details
| Gene Symbol: Nckap5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863012.3 | XP_004863069.1 | 8 - 9 | 3291 | 60 | |
| XM_021243043.1 | XP_021098702.1 | 9 - 10 | 3354 | 60 | ||
| XM_021243044.1 | XP_021098703.1 | 8 - 9 | 3291 | 60 | ||
More Information
Set Membership: |
|

Nckap5l
Genomic Map