Assay Details
| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013070487.2 | XP_012925941.1 | 2 - 3 | 334 | 60 | |
| XM_021240645.1 | XP_021096304.1 | 2 - 3 | 335 | 60 | ||
More Information
Set Membership: |
|

Pcbp4
Genomic Map