Assay Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847258.2 | XP_004847315.1 | 9 - 10 | 1969 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl5| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847258.2 | XP_004847315.1 | 9 - 10 | 1969 | 70 | |
Set Membership: |
|