Assay Details
| Gene Symbol: Pmp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870753.2 | XP_004870810.1 | 4 - 5 | 548 | 62 | |
| XM_004870754.3 | XP_004870811.1 | 4 - 5 | 365 | 62 | ||
More Information
Set Membership: |
|

Pmp22
Genomic Map