Assay Details
| Gene Symbol: Ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861371.2 | XP_004861428.1 | 3 - 4 | 449 | 71 | |
| XM_021241422.1 | XP_021097081.1 | 2 - 3 | 1317 | 71 | ||
More Information
Set Membership: |
|

Ormdl2
Genomic Map