Assay Details
| Gene Symbol: Cdca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836839.2 | XP_004836896.1 | 1 - 2 | 228 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836839.2 transcripts.
Set Membership: |
|

Cdca4
Genomic Map