Assay Details
| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845951.2 | XP_004846008.1 | 3 - 4 | 575 | 64 | |
| XM_021259221.1 | XP_021114880.1 | 3 - 4 | 575 | 64 | ||
More Information
Set Membership: |
|

Rspo3
Genomic Map