Assay Details
| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859628.3 | XP_004859685.2 | 4 - 5 | 450 | 79 | |
| XM_021239947.1 | XP_021095606.1 | 4 - 5 | 450 | 79 | ||
| XM_021239948.1 | XP_021095607.1 | 4 - 5 | 450 | 79 | ||
| XM_021239949.1 | XP_021095608.1 | 4 - 5 | 450 | 79 | ||
More Information
Set Membership: |
|

Snf8
Genomic Map