Assay Details
| Gene Symbol: Supt16h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865047.3 | XP_004865104.1 | 23 - 24 | 3200 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Supt16h| Gene Symbol: Supt16h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865047.3 | XP_004865104.1 | 23 - 24 | 3200 | 65 | |
Set Membership: |
|