Assay Details
| Gene Symbol: Prep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875077.3 | XP_004875134.1 | 14 - 15 | 2086 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prep| Gene Symbol: Prep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875077.3 | XP_004875134.1 | 14 - 15 | 2086 | 70 | |
Set Membership: |
|