Assay Details
| Gene Symbol: Gpx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869239.3 | XP_004869296.1 | 1 - 2 | 182 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpx7| Gene Symbol: Gpx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869239.3 | XP_004869296.1 | 1 - 2 | 182 | 91 | |
Set Membership: |
|