Assay Details
| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849990.3 | XP_004850047.1 | 1 - 2 | 205 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem17| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849990.3 | XP_004850047.1 | 1 - 2 | 205 | 66 | |
Set Membership: |
|