Assay Details
| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841963.3 | XP_004842020.1 | 2 - 3 | 454 | 61 | |
| XM_004841964.3 | XP_004842021.1 | 2 - 3 | 454 | 61 | ||
More Information
Set Membership: |
|

Decr1
Genomic Map