Assay Details
| Gene Symbol: Hcrtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850820.2 | XP_004850877.1 | 6 - 7 | 1259 | 58 | |
| XM_004850821.2 | XP_004850878.1 | 6 - 7 | 1307 | 58 | ||
More Information
Set Membership: |
|

Hcrtr1
Genomic Map