Assay Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869538.2 | XP_004869595.1 | 3 - 4 | 1467 | 84 | |
| XM_021248147.1 | XP_021103806.1 | 2 - 3 | 1362 | 84 | ||
More Information
Set Membership: |
|

Klrg1
Genomic Map