Assay Details
| Gene Symbol: Tmem35b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850915.2 | XP_004850972.1 | 2 - 3 | 330 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem35b| Gene Symbol: Tmem35b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850915.2 | XP_004850972.1 | 2 - 3 | 330 | 80 | |
Set Membership: |
|