Assay Details
| Gene Symbol: Nppc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868058.3 | XP_004868115.1 | 1 - 2 | 183 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nppc| Gene Symbol: Nppc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868058.3 | XP_004868115.1 | 1 - 2 | 183 | 101 | |
Set Membership: |
|