Assay Details
| Gene Symbol: Pitx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867586.2 | XP_004867643.1 | 2 - 3 | 596 | 60 | |
| XM_021246644.1 | XP_021102303.1 | 3 - 4 | 400 | 60 | ||
| XM_021246645.1 | XP_021102304.1 | 2 - 3 | 390 | 60 | ||
More Information
Set Membership: |
|

Pitx2
Genomic Map