Assay Details
| Gene Symbol: Svbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871832.3 | XP_004871889.1 | 2 - 3 | 293 | 79 | |
| XM_004871833.3 | XP_004871890.1 | 2 - 3 | 401 | 79 | ||
More Information
Set Membership: |
|

Svbp
Genomic Map