Assay Details
| Gene Symbol: Bex3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069405.2 | XP_012924859.1 | 2 - 3 | 254 | 90 | |
| XM_013069406.2 | XP_012924860.1 | 2 - 3 | 83 | 90 | ||
| XM_013069412.2 | XP_012924866.1 | 2 - 3 | 226 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013069405.2, XM_013069406.2, XM_013069412.2 transcripts.
Set Membership: |
|

Bex3
Genomic Map