Assay Details
| Gene Symbol: Upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843542.2 | XP_004843599.1 | 3 - 4 | 445 | 57 | |
| XM_004843544.3 | XP_004843601.1 | 2 - 3 | 237 | 57 | ||
| XM_013071693.2 | XP_012927147.1 | 2 - 3 | 586 | 57 | ||
| XM_021257269.1 | XP_021112928.1 | 4 - 5 | 400 | 57 | ||
| XM_021257270.1 | XP_021112929.1 | 3 - 4 | 439 | 57 | ||
| XM_021257271.1 | XP_021112930.1 | 3 - 4 | 301 | 57 | ||
| XM_021257272.1 | XP_021112931.1 | 3 - 4 | 428 | 57 | ||
| XM_021257273.1 | XP_021112932.1 | 2 - 3 | 351 | 57 | ||
More Information
Set Membership: |
|

Upb1
Genomic Map