Assay Details
| Gene Symbol: Galnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835925.2 | XP_004835982.1 | 6 - 7 | 2493 | 66 | |
| XM_021266096.1 | XP_021121755.1 | 6 - 7 | 808 | 66 | ||
More Information
Set Membership: |
|

Galnt5
Genomic Map