Assay Details
| Gene Symbol: Mrpl52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864199.3 | XP_004864256.1 | 3 - 4 | 467 | 95 | |
| XM_004864200.3 | XP_004864257.1 | 3 - 4 | 223 | 95 | ||
| XM_021243910.1 | XP_021099569.1 | 3 - 4 | 517 | 95 | ||
More Information
Set Membership: |
|

Mrpl52
Genomic Map